Orders LinkContactUsLinkComponent
General purpose hydrometers, S50
1,800 - 1,850 g/ml, 0,002 g/ml, 190 mm
Katalog # ALLA3502SC18060HQP
Leverandør:   Alla France
General purpose hydrometers, S50
Katalog # ALLA3502SC18060HQP
Leverandør:  Alla France
General purpose hydrometers, S50
General purpose hydrometers, S50
1,800 - 1,850 g/ml, 0,002 g/ml, 190 mm
Price
522,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
General purpose hydrometers graduated in SG and calibrated at 60 °F (15 °C).
 : ISO 650
Specifikationer
  • Længde:
    190 mm
  • Inddeling:
    0,002 g/ml
  • Område:
    1,800 - 1,850 g/ml