Orders LinkContactUsLinkComponent
Automation tips for Tecan®, Axygen®
Automation tips for Tecan®, Axygen®
EVF-180-R-S, 180 µl, +, +, Transparent, MCA96 / HV96, 50 racks, 96 i hvert
Katalog # 736-0454
Leverandør:   Corning
Automation tips for Tecan®, Axygen®
Katalog # 736-0454
Leverandør:  Corning
Automation tips for Tecan®, Axygen®
Automation tips for Tecan®, Axygen®
EVF-180-R-S, 180 µl, +, +, Transparent, MCA96 / HV96, 50 racks, 96 i hvert
Price
5.680,00 kr
Log ind for tilgængelighed
1 * 4.800 Items
Product Details & Documents
PP, med eller uden filter, sterile eller ikke sterile.
  • Black tips are conductive for liquid level-sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Caution: * Economy packed in cardboard wrapper.
Specifikationer
  • Pakket:
    50 racks, 96 i hvert
  • Steril:
    +
  • Volumen:
    180 µl
  • Seq:
    30
  • Spidstype:
    EVF-180-R-S
  • Spits farge:
    Transparent
  • Til brug med:
    MCA96 / HV96
  • Filter:
    +