Pipette tips, universal, Corning®
Affaset åbning, rund, 100 - 1000 µl, -, Blue, blå, 10 hængslede racks, 100 i hver
: Corning
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
- Beveled orifice helps ensure accurate fluid delivery
- All tips are certified RNase-/DNase-free and non pyrogenic
- Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
- Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
- Farvekode:blå
- Pakket:10 hængslede racks, 100 i hver
- Steril:-
- Volumen:100 - 1000 µl
- Seq:20
- Spidstype:Affaset åbning, rund
- Spits farge:Blue
- Cat. No.:613-5500
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Lav fremstillingspåvirkning