Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør, Corning®

Centrifugerør, Corning®

50 ml, PP, Skruelåg, 15500×g, Conical, 25/rack
Katalog # 525-3414
   Lav fremstillingspåvirkning
Leverandør:   Corning
Centrifugerør, Corning®
Katalog # 525-3414
Leverandør:  Corning
   Lav fremstillingspåvirkning
Centrifugerør, Corning®
Centrifugerør, Corning®

50 ml, PP, Skruelåg, 15500×g, Conical, 25/rack

Price
kr1,420.00
Log ind for tilgængelighed
1 * 300 Items
Product Details & Documents

Corning 50 ml centrifuge tubes made of PP or PET.

  • Conical, with HDPE CentriStar™ or plug seal caps or without cap, sterile
  • Black printed graduations and a large white marking spot
  • Certified non pyrogenic and DNase/RNase-free

The CentriStar™ cap with leakproof design has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.

Bestillingsoplysninger: Skumracks og CentriStar™-låg fås separat.

Emballering: Bulk or in racks.

Specifikationer
  • Lukkestørrelse:
    Skruelåg
  • Pakket:
    25/rack
  • Model:
    Conical
  • Kapacitet:
    50 ml
  • Materiale:
    PP
  • Seq:
    06
  • RCF maks.:
    15500×g
  • Cat. No.:
    525-3414
  • Miljøcertificering:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.