Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for Corning CellSTACK culture chambers

Accessories for Corning CellSTACK culture chambers

Vent cap, polyethylene, 33 mm, 0,2 µm membrane, sterile
Katalog # 734-4166
Leverandør:   Corning
Accessories for Corning CellSTACK culture chambers
Katalog # 734-4166
Leverandør:  Corning
Accessories for Corning CellSTACK culture chambers
Accessories for Corning CellSTACK culture chambers

Vent cap, polyethylene, 33 mm, 0,2 µm membrane, sterile

Price
399,00 kr
Log ind for tilgængelighed
1 * 6 Items
Specifikationer
  • Beskrivelse:
    Vent cap, polyethylene, 33 mm, 0,2 µm membrane, sterile
  • Seq:
    24