pH indicator, phenol red tablets and solutions, Lovibond®
Phenol red tablet, 6,8 – 8,4 pH, Comparator 2000, 100 Tab.
Leverandør: Lovibond
pH indicator, phenol red tablets and solutions, Lovibond®
Katalog # LOVI511750BT
Leverandør: Lovibond
CAS nummer:
Gå til:
Product Details & Documents
Indicator for pH range 6,5 to 8,4.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:100 Tab.
- Beskrivelse:Phenol red tablet
- Til enhed:Comparator 2000
- Område:6,8 – 8,4 pH



