Orders LinkContactUsLinkComponent
Verification standard kit, MultiDirect
Verification standard kit, MultiDirect
Verification standard kit, Multidirect 0/430/530/560/580/610/660 nm, 1 KIT
Katalog # LOVI215650
Leverandør:   Lovibond
Verification standard kit, MultiDirect
Katalog # LOVI215650
Leverandør:  Lovibond
CAS nummer:  
Verification standard kit, MultiDirect
Verification standard kit, MultiDirect
Verification standard kit, Multidirect 0/430/530/560/580/610/660 nm, 1 KIT
CAS nummer:  
Price
kr2,470.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Verification accuracy of photometers and validity of results.
  • Check the performance of photometers over the full measurement range
  • Results can be checked against tolerances provided on the certificate of conformity
Delivery information: Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    Verification standard kit, Multidirect 0/430/530/560/580/610/660 nm