Læselup, easyPOCKET
easyPOCKET, black, 3×, 50×45 mm
Katalog # 630-1037
Leverandør: ESCHENBACH OPTIK
Gå til:
Product Details & Documents
These pocket magnifiers provide magnification and illumination in a credit card format.
- Ultra-flat and lightweight design
- Diffractive aspheric PXM® lightweight lens
- SMD LED illumination, which is switched on when you slide the lens out of its case
- Easy handling - No bulb change required
Leveringsinformation: Supplied with two button cells.
Specifikationer
- Forstørrelse:3×
- Beskrivelse:easyPOCKET, black
- Seq:01
- B×D:50×45 mm