Instrument dishes
5500 ml, 250×350×100 mm
Katalog # BURK4230-2550
Leverandør: BURKLE
Gå til:
Product Details & Documents
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
- Melamine, white
- Tray without lid
Lid available as accessory.
Specifikationer
- Kapacitet:5500 ml
- B×D×H:250×350×100 mm