Orders LinkContactUsLinkComponent
Accessories for SCAT Level Controls
Accessories for SCAT Level Controls
Velcro strip for disc sensor, 2 m
Katalog # 554-2715
Leverandør:   S.C.A.T.
Accessories for SCAT Level Controls
Katalog # 554-2715
Leverandør:  S.C.A.T.
Accessories for SCAT Level Controls
Accessories for SCAT Level Controls
Velcro strip for disc sensor, 2 m
Price
239,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
  • Switching cables
  • Disk sensors
  • Signal cables
  • Power supply units
Specifikationer
  • Beskrivelse:
    Velcro strip for disc sensor, 2 m
  • Seq:
    17