Storage bags
Storage bags, transparent, with ground weld., 0,05 mm, 100×150 mm
Katalog # SEMA2440
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Emballering: Minimum quantity:100 pieces
Specifikationer
- Beskrivelse:Storage bags, transparent, with ground weld.
- Dimensioner:100×150 mm
- Seq:02
- Tykkelse:0,05 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



