Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Blood dilution vials

Blood dilution vials

25,00 ml, 35×56 mm, Yes, Trays of 50, 20 trays per case
Katalog # SIMPV130
Leverandør:   Simport Scientific
Blood dilution vials
Katalog # SIMPV130
Leverandør:  Simport Scientific
Blood dilution vials
Blood dilution vials

25,00 ml, 35×56 mm, Yes, Trays of 50, 20 trays per case

Price
6.300,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents

The dilution vials are made of PS with optically clear parallel sides. The snap cap is spill proof and can be easily removed.

Specifikationer
  • Kasket inkluderet:
    Yes
  • Diameter×H:
    35×56 mm
  • Pakket:
    Trays of 50, 20 trays per case
  • Kapacitet:
    25,00 ml
  • Seq:
    01