Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Push-in stoppers for tubes

gul, 12 mm
  SIMPT401-3Y
Push-in stoppers for tubes
  SIMPT401-3Y
Push-in stoppers for tubes
Push-in stoppers for tubes

gul, 12 mm

 
1.690,00 kr
   
1 * 4.000 Items
 

HDPE. Fit tubes of 15 ml capacity and 17 mm Ø.

  • Impermeable to liquids
  • Can be inserted in tubes and removed again easily with one hand
  • Economical, can also be used as disposable stoppers
 
  • Ø bund:
    12 mm
  • Farve:
    gul
  • Seq:
    09