Storage Bags
150×200 mm
Katalog # 300-0045
Leverandør: Alpha Packaging
Gå til:
Product Details & Documents
Heavy duty storage bags, 500 gauge. Suitable for heat sealing. Clear PE, plain.
Specifikationer
- Dimensioner:150×200 mm
- Seq:01
Heavy duty storage bags, 500 gauge. Suitable for heat sealing. Clear PE, plain.