Weighing boats, PYREX®
3 ml, 70 mm
611-3138
Borosilicate glass.
- Ideal for weighing small quantities of powder
- With hollow stem to facilitate introduction of the powder into narrow neck vessels e.g. volumetric flasks
- Flat base ensures stability on the balance pan
- Kapacitet:3 ml
- Længde:70 mm
- Seq:02