Orders LinkContactUsLinkComponent
Weighing boats, PYREX®
Weighing boats, PYREX®
3 ml, 70 mm
  611-3138
 :   SciLabware
Weighing boats, PYREX®
  611-3138
Weighing boats, PYREX®
Weighing boats, PYREX®
3 ml, 70 mm
2.270,00 kr
1 * 10 Items
Borosilicate glass.
  • Ideal for weighing small quantities of powder
  • With hollow stem to facilitate introduction of the powder into narrow neck vessels e.g. volumetric flasks
  • Flat base ensures stability on the balance pan
  • Kapacitet:
    3 ml
  • Længde:
    70 mm
  • Seq:
    02