Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, suitable only for use with dual block model, 96 well PCR plate, skirted or non-skirted, —
Katalog # JENC357-139
Leverandør: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Katalog # JENC357-139
Leverandør: JENCONS SCIENTIFIC
Gå til:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifikationer
- Ant. huller:—
- Beskrivelse:Interchangeable block, suitable only for use with dual block model
- Seq:18
- Til:96 well PCR plate, skirted or non-skirted