Orders LinkContactUsLinkComponent
Interchangeable blocks for block heaters, digital, SB1
Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, suitable only for use with dual block model, 96 well PCR plate, skirted or non-skirted, —
Katalog # JENC357-139
Leverandør:   JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Katalog # JENC357-139
Leverandør:  JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, suitable only for use with dual block model, 96 well PCR plate, skirted or non-skirted, —
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifikationer
  • Ant. huller:
  • Beskrivelse:
    Interchangeable block, suitable only for use with dual block model
  • Seq:
    18
  • Til:
    96 well PCR plate, skirted or non-skirted