Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 17 mm Ø tube, 12
JENC357-133
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
- Ant. huller:12
- Beskrivelse:Interchangeable block
- Seq:12
- Til:17 mm Ø tube