Orders LinkContactUsLinkComponent
Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 17 mm Ø tube, 12
  JENC357-133
Interchangeable blocks for block heaters, digital, SB1
  JENC357-133
Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 17 mm Ø tube, 12
1 * 1 Items
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
  • Ant. huller:
    12
  • Beskrivelse:
    Interchangeable block
  • Seq:
    12
  • Til:
    17 mm Ø tube