Petriskåle
Uden absorberende pads, 50 mm, 9 mm, Steril, 25 poser med 20 stk.
Katalog # SIMPD210-18A
Leverandør: Simport Scientific
Gå til:
Product Details & Documents
PS, sterile. These 50×9 mm Petri dishes with absorbent pads are ideal for culturing micro-organisms on either agar or broth based media.
- Stackable
- Squared off edges on both the top and bottom provide convenient grips for one handed opening
- Snug fitting top ensures that neither the absorbent pad nor the agar media will dry out during incubation
- Frosted area on top of lid permits labelling
They are designed to accommodate 47 mm diameter membrane filters.
Specifikationer
- Ø:50 mm
- Pakket:25 poser med 20 stk.
- Sterilitet:Steril
- Version:Uden absorberende pads
- Højde:9 mm



