Orders LinkContactUsLinkComponent
Silver seals
Silver seals
Silver seal, for split/splitless injector, 10/pack
Katalog # REST23057
Leverandør:   Restek
Silver seals
Katalog # REST23057
Leverandør:  Restek
Silver seals
Silver seals
Silver seal, for split/splitless injector, 10/pack
Price
kr1,870.00
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Silver seals for Thermo Scientific TRACE, 8000, 8000 TOP and Focus.
Specifikationer
  • Beskrivelse:
    Silver seal, for split/splitless injector, 10/pack
  • Seq:
    01