Silver seals
Silver seal, for split/splitless injector, 10/pack
Katalog # REST23057
Leverandør: Restek
Gå til:
Product Details & Documents
Silver seals for Thermo Scientific TRACE, 8000, 8000 TOP and Focus.
Specifikationer
- Beskrivelse:Silver seal, for split/splitless injector, 10/pack
- Seq:01