0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
Filtre
Filtrerpapir
Kvantitativt filtrerpapir
Quantitative filter papers
Quantitative filter papers
32 mm
512-0586
:
GERBER FUNKE
Quantitative filter papers
512-0586
:
GERBER FUNKE
Quantitative filter papers
32 mm
1.360,00 kr
1 * 1.000 Items
-
+
Størrelse:
32 mm
Lumitester smart, Kikkoman
#: 710-2244
: HYSERVE
pH-teststrimler, MQuant®, Supelco®
#: 1.09535.0001
: Merck
Teststrips, MQuant™, Supelco®
#: 1.10011.0001
: Merck