Orders LinkContactUsLinkComponent
Sediment testers for milk
Sediment testers for milk
Rustfrit stål, 500 ml, SEDILAB
Katalog # 511-0076
Leverandør:   GERBER FUNKE
Sediment testers for milk
Katalog # 511-0076
Leverandør:  GERBER FUNKE
Sediment testers for milk
Sediment testers for milk
Rustfrit stål, 500 ml, SEDILAB
Price
3.590,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
SEDILAB: Manual sediment tester. With clamp for attaching to tables.
SEDILAB-E: Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk. Splash-proof design. Approx. 800 samples per hour. Sharply defined sediment images.
Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk.
Specifikationer
  • Model:
    SEDILAB
  • Kapacitet:
    500 ml
  • Materiale:
    Rustfrit stål