Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Silikoneslanger, PharmaFocus®

25,40 mm, 6,35 mm, 34,93 mm
Silikoneslanger, PharmaFocus®
  228-1178
Silikoneslanger, PharmaFocus®
Silikoneslanger, PharmaFocus®

25,40 mm, 6,35 mm, 34,93 mm

 
10.360,00 kr
   
1 * 15 m
 

Tubing, silicone, platinum cured.

  • More cost-effective silicone tubing that delivers consistent quality and the highest level of assurance
  • Compliant with the requirements of USP, Class VI
  • FDA 21CFR 177-2600/ISO 10093-1/EP 3.1.9

The platinum-cured silicone elastomer used for the PharmaFocus tubing has been designed specifically to fit the needs of the pharmaceutical industry. This silicone undergoes a battery of biocompatibility tests appropriate for pharmaceutical applications, eliminating the costly extended implant studies typically performed on biomedical grade silicones.

Hardness (Shore durometer A): 50

 : Packed double-bagged in boxes. Material certification and lot traceability documents are included in every box.

 
  • Rør udv.Ø:
    34,93 mm
  • Rør int.Ø:
    25,40 mm
  • Vægtykkelse:
    6,35 mm