Sterilin™ Test Tubes
11 ml, 95 mm, 16 mm, Round base plastic tubes, Polystyren, Base tube, Ikke steril
Katalog # SLIN30932
Leverandør: STERILIN
Gå til:
Product Details & Documents
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic round base tubes are manufactured from virgin polystyrene and supplied non sterile. Not available in all geographic regions.
- Convenient alternative to glass tubes
- Suitable for samples ranging from 0,6 to 11 ml
Leveringsinformation: Supplied clean and dust free.
Specifikationer
- Udv.Ø:16 mm
- Sterilitet:Ikke steril
- Model:Base tube
- Kapacitet:11 ml
- Beskrivelse:Round base plastic tubes
- Længde:95 mm
- Materiale:Polystyren
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



