Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumthiosulfat, Supelco®

Natriumthiosulfat, Supelco®

Plastic bottle, 250 g
Katalog # 1.06512.0250
Leverandør:   Merck
Natriumthiosulfat, Supelco®
Katalog # 1.06512.0250
Leverandør:  Merck
CAS nummer:  7772-98-7
Natriumthiosulfat, Supelco®
Natriumthiosulfat, Supelco®

Plastic bottle, 250 g

CAS nummer:  7772-98-7
Price
458,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS nummer:7772-98-7
  • Formel:Na₂S₂O₃
  • MDL nummer:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Merck Index:12,08844
  • Kogende Pt:100 °C (1013 hPa)
  • Smeltende Pt:48 °C
  • Tæthed:1,66 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-867-5
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.