Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rat Recombinant M-CSF (from E. coli)

Rat Recombinant M-CSF (from E. coli)

Katalog # ABCAAB56288-10
Leverandør:   Abcam
Rat Recombinant M-CSF (from E. coli)
Katalog # ABCAAB56288-10
Leverandør:  Abcam
CAS nummer:  
Rat Recombinant M-CSF (from E. coli)
Rat Recombinant M-CSF (from E. coli)
CAS nummer:  
Price
3.370,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant rat M-CSF protein (Active) is a Rat Fragment protein, in the 33 to 186 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Cat. No.:
    ABCAAB56288-10
  • Protein/peptide type:
    Recombinant
  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    M-CSF
  • Protein synonyms:
    Csfm|Csf1|Macrophage colony-stimulating factor 1|CSF-1|MCSF|Proteoglycan macrophage colony-stimulating factor|PG-M-CSF
  • Biologisk aktivitet:
    Standard
  • Renhed:
    >95%
  • Tested applications:
    FuncS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Rat
  • Opbevaring betingelser:
    –80 °C
  • Dyrefri:
    N

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.