PFA inert sample introduction kit supplies
Inert kit for ICP-MS enables analysis of high-purity and HF-containing samples using a PFA-based sample intro system with inert Pt or sapphire-injector torch.
- The O-ring free design and PFA construction material ensure low Background Equivalent Concentrations (BEC) and Detection Limits (DL)
- Added flexibility for organic solvent analysis with an optional connecting pipe with gas port for the addition of O₂/Ar
- Compatible with PFA MicroFlow concentric nebulizer, PFA MicroFlow nebuliser with I-AS probe, PFA concentric 200 μl/min nebulizer and PFA concentric 200 µl/min nebulizer with probe
They manufacture a range of high quality ICP-MS replacements, accessories and consumables including PFA Inert Sample Introduction Kit Supplies. A full range of accessories are available including connector and drain tubes, injector assemblies and torches for 7500 and 7700 ICP-MS systems. To maintain system performance, ensure that consumables are periodically replaced in accordance with recommended maintenance schedules.
A complete kit containing a PFA end cap, PFA spray chamber, connecting tube, demountable quartz torch, torch injector (four options are available: platinum or sapphire with a 2,5 mm or 1,5 mm injector) with PFA base. Easy to install in the standard Peltier-cooled spray chamber mount of the 7700. System uses a simple push-fit torch connector to aid assembly and disconnection
- Beskrivelse:Spray chamber for inert kit, PFA, suits inert sample introduction kit with Agilent 7500/7700/8800 series ICP-MS
- Seq:10