Headspace screw top vials
10,00 ml, 23×46 mm, ravfarvet, No
Katalog # HEWL5188-6538
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
The CombiPAL precision-threaded headspace screw top vials are ideal for applications in the environmental, food and beverage, industrial hygiene, drug analysis, and clinical chemistry industries.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Kasket inkluderet:No
- Diameter×H:23×46 mm
- Kapacitet:10,00 ml
- Farve:ravfarvet
- Seq:01