Inlet liners
Helix liner, double taper, deactivated
Katalog # HEWL5188-5398
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifikationer
- Beskrivelse:Helix liner, double taper, deactivated
- Seq:65