Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to female, 75 mm, I.Ø 0,12 mm, to connect micro valve to detector
Catalog # HEWLG1316-87326
Capillaries for LC systems
Catalog # HEWLG1316-87326
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to female, 75 mm, I.Ø 0,12 mm, to connect micro valve to detector
Price
746,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Beskrivelse:
    Capillaries, non-swaged, male to female, 75 mm, I.Ø 0,12 mm, to connect micro valve to detector
  • Seq:
    36