Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to female, 50 mm, I.Ø 0,12 mm, to connect column to DAD cell
  HEWLG1316-87312
Capillaries for LC systems
  HEWLG1316-87312
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to female, 50 mm, I.Ø 0,12 mm, to connect column to DAD cell
717,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    Capillaries, non-swaged, male to female, 50 mm, I.Ø 0,12 mm, to connect column to DAD cell
  • Seq:
    34