Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
  HEWLG1315-87339
Capillaries for LC systems
  HEWLG1315-87339
Capillaries for LC systems
Capillaries for LC systems
DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
1.000,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
  • Seq:
    92