Capillaries for LC systems
DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
HEWLG1315-87339
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:DAD heat exchanger capillary, 1 end pre-swaged, male to male, 310 mm, I.Ø 0,12 mm
- Seq:92