Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect cell out to MS
Catalog # HEWLG1315-87307
Capillaries for LC systems
Catalog # HEWLG1315-87307
Capillaries for LC systems
Capillaries for LC systems
Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect cell out to MS
Price
784,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Beskrivelse:
    Capillaries, 1 end pre-swaged, male to male, 500 mm, I.Ø 0,12 mm, to connect cell out to MS
  • Seq:
    10