Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
Katalog # HEWLG1312-87303
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1312-87303
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
Price
717,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
  • Seq:
    55