Capillaries for LC systems
Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
Katalog # HEWLG1312-87303
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Capillaries, pre-swaged, male to male, 400 mm, I.Ø 0,17 mm, to connect pump to autosampler
- Seq:55