Capillaries for LC systems
Absorber capillary, 500 μl, pre-swaged, male to male, 1770 mm, I.Ø 0,17 mm
Katalog # HEWLG1312-87300
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
- Beskrivelse:Absorber capillary, 500 μl, pre-swaged, male to male, 1770 mm, I.Ø 0,17 mm
- Seq:01