Orders LinkContactUsLinkComponent
Capillaries for LC systems
Calibration capillary assembly
Katalog # HEWLG1312-67500
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1312-67500
Capillaries for LC systems
Capillaries for LC systems
Calibration capillary assembly
Price
4.150,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Calibration capillary assembly
  • Seq:
    02