Orders LinkContactUsLinkComponent
Convenience kits for inlet systems, QuickPick
Convenience kits for inlet systems, QuickPick
QuickPick purged packed inlet PM kit, with 5 non-stick BTO septa, 1 O-ring, 1 ferrule and 1 disposable glass liner
Katalog # HEWL5188-6498
Leverandør:   AGILENT HEWLETT PACKARD
Convenience kits for inlet systems, QuickPick
Katalog # HEWL5188-6498
Convenience kits for inlet systems, QuickPick
Convenience kits for inlet systems, QuickPick
QuickPick purged packed inlet PM kit, with 5 non-stick BTO septa, 1 O-ring, 1 ferrule and 1 disposable glass liner
Price
321,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    QuickPick purged packed inlet PM kit, with 5 non-stick BTO septa, 1 O-ring, 1 ferrule and 1 disposable glass liner
  • Seq:
    06