Orders LinkContactUsLinkComponent
Convenience kits for inlet systems, QuickPick
Convenience kits for inlet systems, QuickPick
QuickPick split Vent and inlet PM kit with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring, inlet gold seal kit and split vent trap with 2 O-rings
Katalog # HEWL5188-6496
Leverandør:   AGILENT HEWLETT PACKARD
Convenience kits for inlet systems, QuickPick
Katalog # HEWL5188-6496
Convenience kits for inlet systems, QuickPick
Convenience kits for inlet systems, QuickPick
QuickPick split Vent and inlet PM kit with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring, inlet gold seal kit and split vent trap with 2 O-rings
Price
2.030,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    QuickPick split Vent and inlet PM kit with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring, inlet gold seal kit and split vent trap with 2 O-rings
  • Seq:
    04