Convenience kits for inlet systems, QuickPick
QuickPick split Vent and inlet PM kit with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring, inlet gold seal kit and split vent trap with 2 O-rings
Katalog # HEWL5188-6496
Leverandør: AGILENT HEWLETT PACKARD
Convenience kits for inlet systems, QuickPick
Katalog # HEWL5188-6496
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:QuickPick split Vent and inlet PM kit with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring, inlet gold seal kit and split vent trap with 2 O-rings
- Seq:04