Convenience kits for inlet systems, QuickPick
QuickPick splitless inlet PM kit, with 5 non-stick BTO septa, 1 splitless liner, 1 non-stick liner O-ring and inlet gold seal kit
Katalog # HEWL5188-6494
Leverandør: AGILENT HEWLETT PACKARD
Convenience kits for inlet systems, QuickPick
Katalog # HEWL5188-6494
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:QuickPick splitless inlet PM kit, with 5 non-stick BTO septa, 1 splitless liner, 1 non-stick liner O-ring and inlet gold seal kit
- Seq:02