Orders LinkContactUsLinkComponent
Convenience kits for inlet systems, QuickPick
Convenience kits for inlet systems, QuickPick
QuickPick split inlet PM kit, with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring and inlet gold seal kit
Katalog # HEWL5188-6493
Leverandør:   AGILENT HEWLETT PACKARD
Convenience kits for inlet systems, QuickPick
Katalog # HEWL5188-6493
Convenience kits for inlet systems, QuickPick
Convenience kits for inlet systems, QuickPick
QuickPick split inlet PM kit, with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring and inlet gold seal kit
Price
523,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    QuickPick split inlet PM kit, with 5 non-stick BTO septa, 1 split liner, 1 non-stick liner O-ring and inlet gold seal kit
  • Seq:
    01