Orders LinkContactUsLinkComponent
Pipette Boxes, with Lid
Pipette box, for 49 each of 5/3/4" pasteur pipettes, 50×50×160 mm
Katalog # BELC1217-11106
Marketsource
Leverandør:   BELLCO GLASS
Pipette Boxes, with Lid
Katalog # BELC1217-11106
Leverandør:  BELLCO GLASS
Marketsource
Pipette Boxes, with Lid
Pipette Boxes, with Lid
Pipette box, for 49 each of 5/3/4" pasteur pipettes, 50×50×160 mm
Price
472,53 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
  • Stackable
  • Silicone pad on inside of top and bottom protect pipette tips
  • Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
Specifikationer
  • Beskrivelse:
    Pipette box, for 49 each of 5/3/4" pasteur pipettes
  • B×D×H:
    50×50×160 mm