Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bariumnitrat ≥99%

Bariumnitrat ≥99%

Glass bottle for solids, 2.500 g
Katalog # A11305.0E
Oxiderende
Irriterende
Leverandør:   Thermo Fisher Scientific
Bariumnitrat ≥99%
Katalog # A11305.0E
CAS nummer:  10022-31-8
Bariumnitrat ≥99%
Bariumnitrat ≥99%

Glass bottle for solids, 2.500 g

CAS nummer:  10022-31-8
Price
1.440,00 kr
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 10022-31-8 Molecular Formula: BaN2O6 Formula Weight: 261.35 Melting Point: 592° dec. Merck Reference: 14,983 MDL No.: MFCD00003442

  • CAS nummer:10022-31-8
  • Formel:Ba(NO₃)₂
  • MDL nummer:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Merck Index:14,983
  • Smeltende Pt:592 °C (dec.)
  • Tæthed:3,24
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-020-5
  • FN:1446
  • ADR:5.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.