Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Calciumfluorid ≥99%

Glass bottle for solids, 100 g
Katalog # A10913.22
Irriterende
Leverandør:   Thermo Fisher Scientific
Calciumfluorid ≥99%
Katalog # A10913.22
CAS nummer:  7789-75-5
Calciumfluorid ≥99%
Calciumfluorid ≥99%

Glass bottle for solids, 100 g

CAS nummer:  7789-75-5
Price
744,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Refractive Index: 1.434 Odour: Odourless Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

  • CAS nummer:7789-75-5
  • Formel:CaF₂
  • MDL nummer:MFCD00010907
  • Molecular Weight:78.07 g/mol
  • Merck Index:14,1667
  • Kogende Pt:∼2500 °C
  • Smeltende Pt:1403 °C
  • Tæthed:3,18
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-188-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.