Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ac-β-Ala-OH 97%

Ac-β-Ala-OH 97%

5 g
Katalog # H50208.06
Leverandør:   Thermo Fisher Scientific
Ac-β-Ala-OH 97%
Katalog # H50208.06
CAS nummer:  3025-95-4
Ac-β-Ala-OH 97%
Ac-β-Ala-OH 97%

5 g

CAS nummer:  3025-95-4
Price
1.960,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 3025-95-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 77° MDL No.: MFCD00037296

Synonymer:  N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
  • CAS nummer:3025-95-4
  • Formel:C₅H₉NO₃
  • MDL nummer:MFCD00037296
  • Molecular Weight:131.1 g/mol
  • Smeltende Pt:77 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:221-185-6
Specifikationer

Specifikationer

  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.