Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Saponin

Glass bottle for solids, 100 g
Katalog # A18820.22
Irriterende
Leverandør:   Thermo Fisher Scientific
Saponin
Katalog # A18820.22
CAS nummer:  8047-15-2
Saponin
Saponin

Glass bottle for solids, 100 g

CAS nummer:  8047-15-2
Price
1.360,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

CAS No.: 8047-15-2 Physical Form: Powder Appearance: White to pale yellow Melting Point: 158° dec. Merck Reference: 14,8365 MDL No.: MFCD00081981

A glycoside used to permeabilize cellular membranes

  • CAS nummer:8047-15-2
  • MDL nummer:MFCD00081981
  • Merck Index:14,8365
  • Smeltende Pt:158 °C (dec.)
  • Tæthed:1,015…1.02 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-462-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer