Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Metanil gul

Metanil gul

Glass bottle for solids, 250 g
Katalog # A17527.30
Ætsende
Leverandør:   Thermo Fisher Scientific
Metanil gul
Katalog # A17527.30
CAS nummer:  587-98-4
Metanil gul
Metanil gul

Glass bottle for solids, 250 g

CAS nummer:  587-98-4
Price
988,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

CAS No.: 587-98-4 Molecular Formula: C18H14N3NaO3S Formula Weight: 375.38 Odour: Odourless pH: ~10 Beilstein Reference: 3831568 Merck Reference: 14,5928 MDL No.: MFCD00007486

Synonymer:  Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
  • CAS nummer:587-98-4
  • Formel:C₁₈H₁₄N₃NaO₃S
  • MDL nummer:MFCD00007486
  • Molecular Weight:375.38 g/mol
  • Merck Index:14,5928
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-608-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.