Metanil gul
Glass bottle for solids, 50 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 587-98-4 Molecular Formula: C18H14N3NaO3S Formula Weight: 375.38 Odour: Odourless pH: ~10 Beilstein Reference: 3831568 Merck Reference: 14,5928 MDL No.: MFCD00007486
Synonymer: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- CAS nummer:587-98-4
- Formel:C₁₈H₁₄N₃NaO₃S
- MDL nummer:MFCD00007486
- Molecular Weight:375.38 g/mol
- Merck Index:14,5928
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-608-2
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 g
- Cat. No.:A17527.18
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



