L(-)-Carnitine ≥99%
Glass bottle for solids, 50 g
Purity: 99+% CAS No.: 541-15-1 Molecular Formula: C7H15NO3 Formula Weight: 161.20 Melting Point: ca 190° dec. Beilstein Reference: 4292315 Merck Reference: 14,1849 MDL No.: MFCD00038747
: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
- :541-15-1
- :C₇H₁₅NO₃
- :MFCD00038747
- :161.20 g/mol
- :14,1849
- :∼ 190 °C (dec.)
- :Stue temperatur
- :208-768-0
- Pakning:Glass bottle for solids
- Pakn:50 g