Procain, HCl 99%
Glass bottle for solids, 250 g
Purity: 99% CAS No.: 51-05-8 Molecular Formula: C13H20N2O2.HCl Formula Weight: 272.78 Odour: Odourless Melting Point: 155-157° Boiling Point: 195-196°/17mm Merck Reference: 14,7757 UN No.: 2811 MDL No.: MFCD00013000
: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride
- :51-05-8
- :C₁₃H₂₁ClN₂O₂
- :MFCD00013000
- :272.77 g/mol
- :14,7757
- :195…196 °C (17 mmHg)
- :155…157 °C
- :Stue temperatur
- :200-077-2
- :2811
- :6.1,III
- Pakning:Glass bottle for solids
- Pakn:250 g