Orders LinkContactUsLinkComponent
Procain, HCl 99%
Glass bottle for solids, 250 g
  A17485.30
Procain, HCl 99%
  A17485.30
 :  51-05-8
Procain, HCl 99%
Procain, HCl 99%
Glass bottle for solids, 250 g
 :  51-05-8
1.530,00 kr
1 * 250 g
Purity: 99% CAS No.: 51-05-8 Molecular Formula: C13H20N2O2.HCl Formula Weight: 272.78 Odour: Odourless Melting Point: 155-157° Boiling Point: 195-196°/17mm Merck Reference: 14,7757 UN No.: 2811 MDL No.: MFCD00013000
 :  2-Diethylaminoethyl 4-aminobenzoate hydrochloride
  •  :51-05-8
  •  :C₁₃H₂₁ClN₂O₂
  •  :MFCD00013000
  •  :272.77 g/mol
  •  :14,7757
  •  :195…196 °C (17 mmHg)
  •  :155…157 °C
  •  :Stue temperatur
  •  :200-077-2
  •  :2811
  •  :6.1,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g