Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Procain, HCl 99%

Glass bottle for solids, 50 g
Katalog # A17485.18
Giftig
Leverandør:   Thermo Fisher Scientific
Procain, HCl 99%
Katalog # A17485.18
CAS nummer:  51-05-8
Procain, HCl 99%
Procain, HCl 99%

Glass bottle for solids, 50 g

CAS nummer:  51-05-8
Price
423,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 51-05-8 Molecular Formula: C13H20N2O2.HCl Formula Weight: 272.78 Odour: Odourless Melting Point: 155-157° Boiling Point: 195-196°/17mm Merck Reference: 14,7757 UN No.: 2811 MDL No.: MFCD00013000

Synonymer:  2-Diethylaminoethyl 4-aminobenzoate hydrochloride
  • CAS nummer:51-05-8
  • Formel:C₁₃H₂₁ClN₂O₂
  • MDL nummer:MFCD00013000
  • Molecular Weight:272.77 g/mol
  • Merck Index:14,7757
  • Kogende Pt:195…196 °C (17 mmHg)
  • Smeltende Pt:155…157 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-077-2
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.