Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Borsyre ≥98%

Glass bottle for solids, 1.000 g
Katalog # A16624.0B
Sundhed
Leverandør:   Thermo Fisher Scientific
Borsyre ≥98%
Katalog # A16624.0B
CAS nummer:  10043-35-3
Borsyre ≥98%
Borsyre ≥98%

Glass bottle for solids, 1.000 g

CAS nummer:  10043-35-3
Price
537,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 98% CAS No.: 10043-35-3 Molecular Formula: BH3O3 Formula Weight: 61.83 Flash Point: None Odour: Odourless pH: 3.8-4.8 Melting Point: ca 185° dec. Beilstein Reference: 1697939 Merck Reference: 14,1336 MDL No.: MFCD00011337

Synonymer:  Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
  • CAS nummer:10043-35-3
  • Formel:B(OH)₃
  • MDL nummer:MFCD00011337
  • Molecular Weight:61.83 g/mol
  • Merck Index:14,1336
  • Kogende Pt:185 °C (1013 hPa)
  • Smeltende Pt:∼ 185 °C (dec.)
  • Tæthed:1,435
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-139-2
  • NÅ:01-2119486683-25
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer